Assay Details
| Gene Symbol: MYB29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254431.1 | NP_001241360.1 | 2 - 3 | 400 | 65 | |
| GenBank mRNA | AB029274.1 | - | 1 - 2 | 90 | 65 | |
| BT097751.1 | - | 2 - 3 | 400 | 65 | ||
More Information
Set Membership: |
|

MYB29
Genomic Map