Assay Details
| Gene Symbol: SFERH-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250105.1 | NP_001237034.1 | 4 - 5 | 614 | 81 | |
| GenBank mRNA | AB062754.1 | - | 4 - 5 | 614 | 81 | |
| JQ972713.1 | - | 4 - 5 | 529 | 81 | ||
More Information
Set Membership: |
|

SFERH-2
Genomic Map