Assay Details
| Gene Symbol: SFERH-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250105.1 | NP_001237034.1 | 7 - 8 | 809 | 110 | |
| GenBank mRNA | AB062754.1 | - | 7 - 8 | 809 | 110 | |
More Information
Set Membership: |
|

SFERH-2
Genomic Map