Assay Details
Gene Symbol: AG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250192.1 | NP_001237121.1 | 3 - 4 | 385 | 69 | |
GenBank mRNA | AF035671.1 | - | 3 - 4 | 385 | 69 | |
BT097300.1 | - | 3 - 4 | 398 | 69 | ||
BT098436.1 | - | 3 - 4 | 398 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
