Assay Details
| Gene Symbol: GLU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF039851.1 | - | 17 - 18 | 2174 | 103 | |
| AK287031.1 | - | 26 - 27 | 3998 | 103 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GLU| Gene Symbol: GLU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF039851.1 | - | 17 - 18 | 2174 | 103 | |
| AK287031.1 | - | 26 - 27 | 3998 | 103 | ||
Set Membership: |
|