Assay Details
| Gene Symbol: MDH-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF068688.1 | - | 1 - 2 | 126 | 58 | |
| AK286070.1 | - | 3 - 4 | 769 | 58 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MDH-1| Gene Symbol: MDH-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF068688.1 | - | 1 - 2 | 126 | 58 | |
| AK286070.1 | - | 3 - 4 | 769 | 58 | ||
Set Membership: |
|