Assay Details
| Gene Symbol: MDH-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF068688.1 | - | 2 - 3 | 191 | 101 | |
| AK286070.1 | - | 4 - 5 | 834 | 101 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MDH-1| Gene Symbol: MDH-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF068688.1 | - | 2 - 3 | 191 | 101 | |
| AK286070.1 | - | 4 - 5 | 834 | 101 | ||
Set Membership: |
|