Assay Details
| Gene Symbol: LOC547948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249537.1 | NP_001236466.1 | 9 - 10 | 1410 | 88 | |
| GenBank mRNA | AF118785.1 | - | 9 - 10 | 1410 | 88 | |
More Information
Set Membership: |
|

LOC547948
Genomic Map