Assay Details
| Gene Symbol: GSH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF128453.1 | - | 8 - 9 | 1029 | 71 | |
| AK286212.1 | - | 8 - 9 | 1033 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GSH1| Gene Symbol: GSH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF128453.1 | - | 8 - 9 | 1029 | 71 | |
| AK286212.1 | - | 8 - 9 | 1033 | 71 | ||
Set Membership: |
|