Assay Details
| Gene Symbol: GRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251444.1 | NP_001238373.1 | 1 - 2 | 108 | 61 | |
| GenBank mRNA | AF169205.1 | - | 1 - 2 | 108 | 61 | |
More Information
Set Membership: |
|

GRP
Genomic Map