Assay Details
| Gene Symbol: SCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251556.1 | NP_001238485.1 | 6 - 7 | 2917 | 143 | |
| GenBank mRNA | AF195028.1 | - | 6 - 7 | 2917 | 143 | |
More Information
Set Membership: |
|

SCA1
Genomic Map