Assay Details
| Gene Symbol: SCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250255.1 | NP_001237184.1 | 5 - 6 | 2793 | 68 | |
| GenBank mRNA | AF195029.1 | - | 5 - 6 | 2793 | 68 | |
More Information
Set Membership: |
|

SCA2
Genomic Map