Assay Details
Gene Symbol: CHIA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251632.1 | NP_001238561.1 | 1 - 2 | 458 | 64 | |
GenBank mRNA | AF202731.1 | - | 1 - 2 | 458 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
