Assay Details
| Gene Symbol: PCS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248647.1 | NP_001235576.1 | 1 - 2 | 218 | 72 | |
| GenBank mRNA | AF411075.1 | - | 1 - 2 | 218 | 72 | |
More Information
Set Membership: |
|

PCS1
Genomic Map