Assay Details
| Gene Symbol: CA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249275.1 | NP_001236204.1 | 8 - 9 | 727 | 107 | |
| GenBank mRNA | AJ239132.1 | - | 8 - 9 | 727 | 107 | |
| BT097689.1 | - | 8 - 9 | 745 | 107 | ||
More Information
Set Membership: |
|

CA1
Genomic Map