Assay Details
| Gene Symbol: AT-1SNBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249331.1 | NP_001236260.1 | 1 - 2 | 990 | 95 | |
| GenBank mRNA | AJ319868.1 | - | 1 - 2 | 990 | 95 | |
More Information
Set Membership: |
|

AT-1SNBP
Genomic Map