Assay Details
| Gene Symbol: PUR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251130.1 | NP_001238059.1 | 4 - 5 | 948 | 88 | |
| GenBank mRNA | AY321444.1 | - | 4 - 5 | 948 | 88 | |
| X96865.1 | - | 4 - 5 | 913 | 88 | ||
More Information
Set Membership: |
|

PUR3
Genomic Map