Assay Details
| Gene Symbol: GAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254052.1 | NP_001240981.1 | 2 - 3 | 288 | 79 | |
| GenBank mRNA | AB240966.1 | - | 2 - 3 | 288 | 79 | |
| AK285497.1 | - | 2 - 3 | 349 | 79 | ||
| AY323131.1 | - | 1 - 2 | 130 | 79 | ||
More Information
Set Membership: |
|

GAD
Genomic Map