Assay Details
| Gene Symbol: GAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254052.1 | NP_001240981.1 | 4 - 5 | 704 | 104 | |
| GenBank mRNA | AB240966.1 | - | 4 - 5 | 704 | 104 | |
| AK285497.1 | - | 4 - 5 | 765 | 104 | ||
| AY323131.1 | - | 3 - 4 | 546 | 104 | ||
More Information
Set Membership: |
|

GAD
Genomic Map