Assay Details
| Gene Symbol: AGL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250104.1 | NP_001237033.1 | 5 - 6 | 698 | 85 | |
| GenBank mRNA | AY370659.1 | - | 5 - 6 | 698 | 85 | |
More Information
Set Membership: |
|

AGL15
Genomic Map