Assay Details
| Gene Symbol: LOC547742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250254.1 | NP_001237183.1 | 6 - 7 | 926 | 93 | |
| GenBank mRNA | AY439100.1 | - | 6 - 7 | 926 | 93 | |
More Information
Set Membership: |
|

LOC547742
Genomic Map