Assay Details
| Gene Symbol: GK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250374.1 | NP_001237303.1 | 1 - 2 | 358 | 65 | |
| GenBank mRNA | AY492004.1 | - | 1 - 2 | 358 | 65 | |
More Information
Set Membership: |
|

GK
Genomic Map