Assay Details
| Gene Symbol: P5CS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251224.1 | NP_001238153.1 | 9 - 10 | 1176 | 110 | |
| GenBank mRNA | AY492005.1 | - | 9 - 10 | 1176 | 110 | |
| FM999729.1 | - | 9 - 10 | 1176 | 110 | ||
| FN564571.1 | - | 9 - 10 | 1176 | 110 | ||
More Information
Set Membership: |
|

P5CS
Genomic Map