Assay Details
| Gene Symbol: F3H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249868.2 | NP_001236797.1 | 1 - 2 | 380 | 86 | |
| GenBank mRNA | AF198451.1 | - | 1 - 2 | 431 | 86 | |
| AY595420.1 | - | 1 - 2 | 431 | 86 | ||
| AY669324.1 | - | 1 - 2 | 423 | 86 | ||
| BT096691.1 | - | 1 - 2 | 382 | 86 | ||
| EF100870.1 | - | 424 | 85 | |||
| EF100871.1 | - | 1 - 2 | 425 | 86 | ||
| FJ770474.1 | - | 1 - 2 | 372 | 86 | ||
More Information
Set Membership: |
|

F3H
Genomic Map