Assay Details
| Gene Symbol: F3H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249868.2 | NP_001236797.1 | 2 - 3 | 814 | 71 | |
| GenBank mRNA | AF198451.1 | - | 2 - 3 | 865 | 71 | |
| AY595420.1 | - | 2 - 3 | 865 | 71 | ||
| AY669324.1 | - | 2 - 3 | 857 | 71 | ||
| BT096691.1 | - | 2 - 3 | 816 | 71 | ||
| EF100871.1 | - | 2 - 3 | 859 | 71 | ||
| FJ770474.1 | - | 2 - 3 | 806 | 71 | ||
More Information
Set Membership: |
|

F3H
Genomic Map