Assay Details
| Gene Symbol: AUX28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249764.1 | NP_001236693.1 | 2 - 3 | 1000 | 96 | |
| GenBank mRNA | AK285945.1 | - | 2 - 3 | 490 | 96 | |
| J03919.1 | - | 2 - 3 | 1000 | 96 | ||
More Information
Set Membership: |
|

AUX28
Genomic Map