Assay Details
| Gene Symbol: AUX28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249764.1 | NP_001236693.1 | 3 - 4 | 1140 | 61 | |
| GenBank mRNA | AK285945.1 | - | 3 - 4 | 630 | 61 | |
| BT097202.1 | - | 3 - 4 | 812 | 61 | ||
| DQ191055.1 | - | 2 - 3 | 290 | 61 | ||
| J03919.1 | - | 3 - 4 | 1140 | 61 | ||
| Gene Symbol: LOC100810013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK285198.1 | - | 3 - 4 | 648 | 61 | |
More Information
Set Membership: |
|

AUX28
Genomic Map