Assay Details
| Gene Symbol: GR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251077.1 | NP_001238006.1 | 3 - 4 | 897 | 115 | |
| GenBank mRNA | L11632.1 | - | 3 - 4 | 897 | 115 | |
| Gene Symbol: LOC100810005 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK286768.1 | - | 3 - 4 | 875 | 115 | |
More Information
Set Membership: |
|

GR
Genomic Map