Assay Details
| Gene Symbol: LOC547983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250026.2 | NP_001236955.1 | 13 - 14 | 1924 | 91 | |
| GenBank mRNA | L27265.1 | - | 13 - 14 | 1924 | 91 | |
More Information
Set Membership: |
|

LOC547983
Genomic Map