Assay Details
| Gene Symbol: LOC547983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250026.2 | NP_001236955.1 | 15 - 16 | 2217 | 99 | |
| GenBank mRNA | AK245999.1 | - | 9 - 10 | 1280 | 99 | |
| L27265.1 | - | 15 - 16 | 2217 | 99 | ||
More Information
Set Membership: |
|

LOC547983
Genomic Map