Assay Details
| Gene Symbol: LOC547983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250026.2 | NP_001236955.1 | 8 - 9 | 1164 | 72 | |
| GenBank mRNA | AK245999.1 | - | 4 - 5 | 553 | 72 | |
| L27265.1 | - | 8 - 9 | 1164 | 72 | ||
| Gene Symbol: LOC100778348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255386.1 | NP_001242315.1 | 8 - 9 | 1214 | 72 | |
| GenBank mRNA | L29770.1 | - | 8 - 9 | 1214 | 72 | |
More Information
Set Membership: |
|

LOC547983
Genomic Map