Assay Details
| Gene Symbol: TBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251273.1 | NP_001238202.1 | 2 - 3 | 249 | 94 | |
| GenBank mRNA | L28002.1 | - | 2 - 3 | 249 | 94 | |
More Information
Set Membership: |
|

TBP1
Genomic Map