Assay Details
| Gene Symbol: PHYA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251357.1 | NP_001238286.1 | 2 - 3 | 2907 | 100 | |
| GenBank mRNA | EU428747.2 | - | 2 - 3 | 2904 | 100 | |
| L34844.1 | - | 2 - 3 | 2907 | 100 | ||
More Information
Set Membership: |
|

PHYA
Genomic Map