Assay Details
| Gene Symbol: DHPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251370.1 | NP_001238299.1 | 1 - 2 | 76 | 73 | |
| GenBank mRNA | L36436.1 | - | 1 - 2 | 76 | 73 | |
More Information
Set Membership: |
|

DHPS1
Genomic Map