Assay Details
| Gene Symbol: LOC547825 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251588.2 | NP_001238517.2 | 2 - 3 | 841 | 109 | |
| GenBank mRNA | M64987.1 | - | 2 - 3 | 843 | 109 | |
More Information
Set Membership: |
|

LOC547825
Genomic Map