Assay Details
| Gene Symbol: LOC547825 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251588.2 | NP_001238517.2 | 3 - 4 | 993 | 78 | |
| GenBank mRNA | M64987.1 | - | 3 - 4 | 995 | 78 | |
| Gene Symbol: LOC100806678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255033.1 | NP_001241962.1 | 3 - 4 | 861 | 78 | |
| GenBank mRNA | AY247754.1 | - | 3 - 4 | 861 | 78 | |
More Information
Set Membership: |
|

LOC547825
Genomic Map