Assay Details
| Gene Symbol: GS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251602.1 | NP_001238531.1 | 6 - 7 | 542 | 64 | |
| GenBank mRNA | AF301590.1 | - | 6 - 7 | 527 | 64 | |
| AK244891.1 | - | 6 - 7 | 561 | 64 | ||
| AY920930.1 | - | 300 | 64 | |||
| BT091697.1 | - | 6 - 7 | 562 | 64 | ||
| BT097849.1 | - | 6 - 7 | 562 | 64 | ||
| S46513.1 | - | 6 - 7 | 542 | 64 | ||
More Information
Set Membership: |
|

GS
Genomic Map