Assay Details
| Gene Symbol: HEMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251808.1 | NP_001238737.1 | 10 - 11 | 1009 | 75 | |
| GenBank mRNA | AK243972.1 | - | 10 - 11 | 1018 | 75 | |
| BT093896.1 | - | 3 - 4 | 420 | 75 | ||
| U04525.1 | - | 10 - 11 | 1009 | 75 | ||
More Information
Set Membership: |
|

HEMB
Genomic Map