Assay Details
| Gene Symbol: HEMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251808.1 | NP_001238737.1 | 6 - 7 | 666 | 103 | |
| GenBank mRNA | AK243972.1 | - | 6 - 7 | 675 | 103 | |
| AK285742.1 | - | 6 - 7 | 722 | 103 | ||
| U04525.1 | - | 6 - 7 | 666 | 103 | ||
More Information
Set Membership: |
|

HEMB
Genomic Map