Assay Details
| Gene Symbol: LOC547845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248093.1 | NP_001235022.1 | 9 - 10 | 755 | 84 | |
| GenBank mRNA | U12926.1 | - | 9 - 10 | 755 | 84 | |
More Information
Set Membership: |
|

LOC547845
Genomic Map