Assay Details
| Gene Symbol: CDC48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248170.1 | NP_001235099.1 | 6 - 7 | 1533 | 83 | |
| GenBank mRNA | AK286854.1 | - | 2878 | 83 | ||
| DQ359725.1 | - | 6 - 7 | 1533 | 83 | ||
| U20213.1 | - | 6 - 7 | 1553 | 83 | ||
More Information
Set Membership: |
|

CDC48
Genomic Map