Assay Details
| Gene Symbol: RAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251238.1 | NP_001238167.1 | 3 - 4 | 364 | 70 | |
| GenBank mRNA | U32185.1 | - | 3 - 4 | 364 | 70 | |
More Information
Set Membership: |
|

RAB2
Genomic Map