Assay Details
| Gene Symbol: LOC548075 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251496.1 | NP_001238425.1 | 6 - 7 | 437 | 102 | |
| GenBank mRNA | U66836.1 | - | 6 - 7 | 437 | 102 | |
More Information
Set Membership: |
|

LOC548075
Genomic Map