Assay Details
| Gene Symbol: LOC548075 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251496.1 | NP_001238425.1 | 8 - 9 | 527 | 65 | |
| GenBank mRNA | U66836.1 | - | 8 - 9 | 527 | 65 | |
More Information
Set Membership: |
|

LOC548075
Genomic Map