Assay Details
| Gene Symbol: GF14D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250136.1 | NP_001237065.1 | 1 - 2 | 84 | 80 | |
| GenBank mRNA | AK285774.1 | - | 1 - 2 | 150 | 80 | |
| U70536.1 | - | 1 - 2 | 84 | 80 | ||
More Information
Set Membership: |
|

GF14D
Genomic Map