Assay Details
| Gene Symbol: GS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF301590.1 | - | 12 | 1307 | 106 | |
| BT097849.1 | - | 12 | 1342 | 106 | ||
| X68598.1 | - | 1 | 249 | 105 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GS| Gene Symbol: GS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF301590.1 | - | 12 | 1307 | 106 | |
| BT097849.1 | - | 12 | 1342 | 106 | ||
| X68598.1 | - | 1 | 249 | 105 | ||
Set Membership: |
|