Assay Details
Gene Symbol: BMY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249318.1 | NP_001236247.1 | 1 - 2 | 170 | 100 | |
GenBank mRNA | AB004271.1 | - | 1 - 2 | 167 | 100 | |
AB189843.1 | - | 1 - 2 | 67 | 100 | ||
AB189916.1 | - | 1 - 2 | 64 | 100 | ||
AK243900.1 | - | 1 - 2 | 171 | 100 | ||
AK285807.1 | - | 1 - 2 | 171 | 100 | ||
D50866.1 | - | 1 - 2 | 135 | 100 | ||
DQ114422.1 | - | 1 - 2 | 61 | 100 | ||
DQ114423.1 | - | 1 - 2 | 61 | 100 | ||
M92090.1 | - | 1 - 2 | 162 | 100 | ||
X71419.1 | - | 1 - 2 | 170 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
