Assay Details
| Gene Symbol: HOP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249332.1 | NP_001236261.1 | 4 - 5 | 1523 | 89 | |
| GenBank mRNA | X79770.1 | - | 4 - 5 | 1523 | 89 | |
More Information
Set Membership: |
|

HOP1
Genomic Map