Assay Details
| Gene Symbol: CRY1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248276.1 | NP_001235205.1 | 1 - 2 | 343 | 80 | |
| GenBank mRNA | AB498928.1 | - | 1 - 2 | 343 | 80 | |
| DQ401046.1 | - | 1 - 2 | 317 | 80 | ||
More Information
Set Membership: |
|

CRY1a
Genomic Map