Assay Details
| Gene Symbol: MYB55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BT098318.1 | - | 2 - 3 | 326 | 106 | |
| DQ822885.1 | - | 2 - 3 | 286 | 106 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYB55| Gene Symbol: MYB55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BT098318.1 | - | 2 - 3 | 326 | 106 | |
| DQ822885.1 | - | 2 - 3 | 286 | 106 | ||
Set Membership: |
|